Isopropyl 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzenesulfonate 95%

Supplier: Ambeed
Warning

A1619492-1G A1619492-250MG A1619492-25G A1619492-5G
76668-814EA 140.04 USD
76668-814 76668-816 76668-818 76668-820
Isopropyl 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzenesulfonate 95%
Isopropyl 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzenesulfonate

Precaución: for Research Use Only

Formula: C₁₅H₂₃BO₅S
MW: 326.22 g/mol
CAS Number: 1324003-62-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR