N-(2-Bromo-5-chlorobenzyl)propan-2-amine 95%
Supplier: Ambeed
|
Warning
|
A1618336-100MG
A1618336-1G
A1618336-250MG
76668-784EA
102
USD
76668-784
76668-786
76668-788
N-(2-Bromo-5-chlorobenzyl)propan-2-amine 95%
N-(2-Bromo-5-chlorobenzyl)propan-2-amine
Precaución: for Research Use Only
Formula:
C₁₀H₁₃BrClN MW: 262.57 g/mol |
MDL Number:
MFCD21705590 CAS Number: 1500242-27-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...