N-(tert-Butyl)-2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide 95%

Supplier: Ambeed
Warning

A1618332-1G A1618332-250MG A1618332-5G
76668-778EA 91 USD
76668-778 76668-780 76668-782
N-(tert-Butyl)-2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide 95%
N-(tert-Butyl)-2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide

Precaución: for Research Use Only

Formula: C₁₇H₂₅BFNO₃
CAS Number: 2828445-61-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR