2-Ethoxy-1-fluoro-4-methylbenzene 95%

Supplier: Ambeed
Warning

A1618232-100MG A1618232-1G A1618232-250MG
76668-766EA 47.75 USD
76668-766 76668-768 76668-770
2-Ethoxy-1-fluoro-4-methylbenzene 95%
2-Ethoxy-1-fluoro-4-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₁FO
MW: 154.18 g/mol
MDL Number: MFCD20219369
CAS Number: 1486959-73-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR