N,N-Diisopropyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide 95%

Supplier: Ambeed
Warning

A1618100-1G A1618100-250MG A1618100-5G
76668-760EA 42 USD
76668-760 76668-762 76668-764
N,N-Diisopropyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide 95%
N,N-Diisopropyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide

Precaución: for Research Use Only

Formula: C₁₉H₃₀BNO₃
MW: 331.25 g/mol
CAS Number: 1509963-29-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR