3-(2-Methoxyethoxy)pyridine 95%

Supplier: Ambeed
Warning

A1618068-100MG A1618068-1G A1618068-250MG
76668-754EA 5 USD
76668-754 76668-756 76668-758
3-(2-Methoxyethoxy)pyridine 95%
3-(2-Methoxyethoxy)pyridine

Precaución: for Research Use Only

Formula: C₈H₁₁NO₂
MDL Number: MFCD18802515
CAS Number: 98684-29-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR