(9-Oxo-9H-fluoren-2-yl)boronic acid 95%

Supplier: Ambeed
Warning

A1618029-1G A1618029-250MG A1618029-5G
76668-748EA 72 USD
76668-748 76668-750 76668-752
(9-Oxo-9H-fluoren-2-yl)boronic acid 95%
(9-Oxo-9H-fluoren-2-yl)boronic acid

Precaución: for Research Use Only

Formula: C₁₃H₉BO₃
MDL Number: MFCD26142008
CAS Number: 2036064-15-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR