2-(3,5-Difluoro-4-propoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%

Supplier: Ambeed
Warning

A1617728-1G A1617728-250MG A1617728-5G
76668-722EA 125.19 USD
76668-722 76668-724 76668-726
2-(3,5-Difluoro-4-propoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%
2-(3,5-Difluoro-4-propoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₅H₂₁BF₂O₃
MW: 298.13 g/mol
MDL Number: MFCD33403760
CAS Number: 2828445-55-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR