1-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)piperazine dihydrochloride 95%

Supplier: Ambeed
Warning

A1617537-1G A1617537-250MG A1617537-5G
76668-704EA 106.09 USD
76668-704 76668-706 76668-708
1-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)piperazine dihydrochloride 95%
1-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)piperazine dihydrochloride

Precaución: for Research Use Only

Formula: C₁₇H₂₉BCl₂N₂O₂
MW: 375.14 g/mol
CAS Number: 2828445-30-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR