2-(2,3-Difluoro-6-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98%

Supplier: Ambeed
Warning

A1616396-1G A1616396-250MG A1616396-5G
76668-680EA 97.61 USD
76668-680 76668-682 76668-684
2-(2,3-Difluoro-6-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98%
2-(2,3-Difluoro-6-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₃H₁₇BF₂O₃
MW: 270.08 g/mol
MDL Number: MFCD24039571
CAS Number: 2737271-23-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR