Dimethyl 2-(bromomethyl)isophthalate 98%

Supplier: Ambeed
Danger

A1616367-1G A1616367-250MG A1616367-5G
76668-674EA 189 USD
76668-674 76668-676 76668-678
Dimethyl 2-(bromomethyl)isophthalate 98%
Dimethyl 2-(bromomethyl)isophthalate

Precaución: for Research Use Only

Formula: C₁₁H₁₁BrO₄
MDL Number: MFCD34563920
CAS Number: 16281-93-9
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR