Methyl 2,6-difluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate 95%
Supplier: Ambeed
|
Warning
|
A1604332-100MG
A1604332-1G
A1604332-250MG
76668-654EA
36.08
USD
76668-654
76668-656
76668-658
Methyl 2,6-difluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate 95%
Methyl 2,6-difluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
Precaución: for Research Use Only
Formula:
C₁₄H₁₇BF₂O₄ MW: 298.09 g/mol |
CAS Number:
2828445-13-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...