1-((4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)sulfonyl)azetidine 95%
Supplier: Ambeed
|
Warning
|
A1604328-1G
A1604328-250MG
A1604328-5G
76668-648EA
160.2
USD
76668-648
76668-650
76668-652
1-((4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)sulfonyl)azetidine 95%
1-((4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)sulfonyl)azetidine
Precaución: for Research Use Only
Formula:
C₁₅H₂₂BNO₄S MW: 323.21 g/mol |
CAS Number:
2781935-67-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...