(3-(Ethoxycarbonyl)-1-trityl-1H-pyrazol-4-yl)boronic acid 95%

Supplier: Ambeed
Warning

A1603413-100MG A1603413-1G A1603413-250MG
76668-616EA 36.08 USD
76668-616 76668-618 76668-620
(3-(Ethoxycarbonyl)-1-trityl-1H-pyrazol-4-yl)boronic acid 95%
(3-(Ethoxycarbonyl)-1-trityl-1H-pyrazol-4-yl)boronic acid

Precaución: for Research Use Only

Formula: C₂₅H₂₃BN₂O₄
MW: 426.27 g/mol
CAS Number: 2828444-94-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR