(4-(4-(tert-Butoxycarbonyl)piperazin-1-yl)-3-nitrophenyl)boronic acid 95%

Supplier: Ambeed
Warning

A1600236-100MG A1600236-1G A1600236-250MG
76668-598EA 39 USD
76668-598 76668-600 76668-602
(4-(4-(tert-Butoxycarbonyl)piperazin-1-yl)-3-nitrophenyl)boronic acid 95%
(4-(4-(tert-Butoxycarbonyl)piperazin-1-yl)-3-nitrophenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₅H₂₂BN₃O₆
CAS Number: 2828444-20-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR