2-(3-(4-Fluorophenoxy)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%
Supplier: Ambeed
|
Warning
|
A1600086-100MG
A1600086-1G
A1600086-250MG
76668-592EA
36.08
USD
76668-592
76668-594
76668-596
2-(3-(4-Fluorophenoxy)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%
2-(3-(4-Fluorophenoxy)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Precaución: for Research Use Only
Formula:
C₁₈H₂₀BFO₃ MW: 314.15 g/mol |
MDL Number:
MFCD22683473 CAS Number: 1367127-80-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...