4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzene-1,2-diol 95%

Supplier: Ambeed
Warning

A1598752-100MG A1598752-1G A1598752-250MG
76668-586EA 89 USD
76668-586 76668-588 76668-590
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzene-1,2-diol 95%
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzene-1,2-diol

Precaución: for Research Use Only

Formula: C₁₂H₁₇BO₄
MW: 236.07 g/mol
MDL Number: MFCD28356376
CAS Number: 2095372-76-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR