(4-(1-Methoxy-2-methyl-1-oxopropan-2-yl)phenyl)boronic acid 95%

Supplier: Ambeed
Warning

A1581451-100MG A1581451-1G A1581451-250MG
76668-580EA 37 USD
76668-580 76668-582 76668-584
(4-(1-Methoxy-2-methyl-1-oxopropan-2-yl)phenyl)boronic acid 95%
(4-(1-Methoxy-2-methyl-1-oxopropan-2-yl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₅BO₄
MW: 222.04 g/mol
CAS Number: 1388233-62-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR