N1,N3-Dibutyl-N1,N3-dimethylmalonamide 95%

Supplier: Ambeed
Warning

A1547263-5G A1547263-1G A1547263-250MG
76668-552EA 540 USD
76668-552 77876-253 77876-254
N1,N3-Dibutyl-N1,N3-dimethylmalonamide 95%
N1,N3-Dibutyl-N1,N3-dimethylmalonamide

Precaución: for Research Use Only

Formula: C₁₃H₂₆N₂O₂
MW: 242.35 g/mol
MDL Number: MFCD00800937
CAS Number: 91914-98-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR