(2-(Methoxycarbonyl)chroman-6-yl)boronic acid 97%

Supplier: Ambeed
Warning

A1546912-100MG A1546912-1G A1546912-250MG
76668-538EA 39 USD
76668-538 76668-540 76668-542
(2-(Methoxycarbonyl)chroman-6-yl)boronic acid 97%
(2-(Methoxycarbonyl)chroman-6-yl)boronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₃BO₅
MW: 236.02 g/mol
CAS Number: 2828444-05-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR