(3-Chloro-4-fluoro-5-(methoxycarbonyl)phenyl)boronic acid 95+%

Supplier: Ambeed
Warning

A1546910-100MG A1546910-1G A1546910-250MG
76668-532EA 37.14 USD
76668-532 76668-534 76668-536
(3-Chloro-4-fluoro-5-(methoxycarbonyl)phenyl)boronic acid 95+%
(3-Chloro-4-fluoro-5-(methoxycarbonyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₇BClFO₄
MW: 232.40 g/mol
CAS Number: 2828444-03-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR