(3,4-Dichloro-5-hydroxyphenyl)boronic acid ≥98%

Supplier: Ambeed
Warning

A1546872-100MG A1546872-1G A1546872-250MG
76668-526EA 36.08 USD
76668-526 76668-528 76668-530
(3,4-Dichloro-5-hydroxyphenyl)boronic acid ≥98%
(3,4-Dichloro-5-hydroxyphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₆H₅BCl₂O₃
MW: 206.81 g/mol
MDL Number: MFCD28786644
CAS Number: 1701449-88-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR