5-Bromo-2-((2-chlorobenzyl)oxy)-1,3-dimethylbenzene 95%

Supplier: Ambeed
Warning

A1544835-100MG A1544835-1G A1544835-250MG
76668-508EA 39 USD
76668-508 76668-510 76668-512
5-Bromo-2-((2-chlorobenzyl)oxy)-1,3-dimethylbenzene 95%
5-Bromo-2-((2-chlorobenzyl)oxy)-1,3-dimethylbenzene

Precaución: for Research Use Only

Formula: C₁₅H₁₄BrClO
MDL Number: MFCD32065075
CAS Number: 2469394-16-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR