N-(2-Chloroethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide 95%

Supplier: Ambeed
Warning

A1544300-1G A1544300-250MG A1544300-5G
76668-482EA 66 USD
76668-482 76668-484 76668-486
N-(2-Chloroethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide 95%
N-(2-Chloroethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide

Precaución: for Research Use Only

Formula: C₁₅H₂₁BClNO₃
CAS Number: 2828443-95-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR