N-Cyclohexyl-N-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide 95%

Supplier: Ambeed
Warning

A1543220-100MG A1543220-1G A1543220-250MG A1543220-5G
76668-440EA 36.08 USD
76668-440 76668-442 76668-444 76668-446
N-Cyclohexyl-N-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide 95%
N-Cyclohexyl-N-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide

Precaución: for Research Use Only

Formula: C₂₀H₃₀BNO₃
MW: 343.26 g/mol
CAS Number: 2828443-89-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR