Methyl 2,4-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate 95+%

Supplier: Ambeed
Warning

A1543055-100MG A1543055-1G A1543055-250MG
76668-408EA 36.08 USD
76668-408 76668-410 76668-412
Methyl 2,4-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate 95+%
Methyl 2,4-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

Precaución: for Research Use Only

Formula: C₂₀H₃₀B₂O₆
MW: 388.07 g/mol
CAS Number: 1417036-37-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR