2-(4'-Isopropoxy-[1,1'-biphenyl]-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98%

Supplier: Ambeed
Warning

A1542831-100MG A1542831-1G A1542831-250MG
76668-384EA 37.14 USD
76668-384 76668-386 76668-388
2-(4'-Isopropoxy-[1,1'-biphenyl]-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98%
2-(4'-Isopropoxy-[1,1'-biphenyl]-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₂₁H₂₇BO₃
MW: 338.24 g/mol
MDL Number: MFCD34705981
CAS Number: 2828440-21-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR