N-Cyclopentyl-2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide 95%

Supplier: Ambeed
Warning

A1541783-100MG A1541783-1G A1541783-250MG
76668-372EA 39 USD
76668-372 76668-374 76668-376
N-Cyclopentyl-2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide 95%
N-Cyclopentyl-2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide

Precaución: for Research Use Only

Formula: C₁₈H₂₅BFNO₃
CAS Number: 2828440-19-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR