2-(4-((4-Chlorobenzyl)oxy)-3,5-dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97%
Supplier: Ambeed
|
Warning
|
A1541435-100MG
A1541435-1G
A1541435-250MG
76668-344EA
35.01
USD
76668-344
76668-346
76668-348
2-(4-((4-Chlorobenzyl)oxy)-3,5-dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97%
2-(4-((4-Chlorobenzyl)oxy)-3,5-dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Precaución: for Research Use Only
Formula:
C₂₁H₂₆BClO₃ MW: 372.69 g/mol |
CAS Number:
2828440-16-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...