(4-Chloro-3-(methoxycarbonyl)-5-methylphenyl)boronic acid 97%

Supplier: Ambeed
Warning

A1541260-100MG A1541260-1G A1541260-250MG
76668-338EA 19 USD
76668-338 76668-340 76668-342
(4-Chloro-3-(methoxycarbonyl)-5-methylphenyl)boronic acid 97%
(4-Chloro-3-(methoxycarbonyl)-5-methylphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₁₀BClO₄
MW: 228.43 g/mol
CAS Number: 2828440-15-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR