2-(2-Fluoro-3-isobutoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%

Supplier: Ambeed
Warning

A1541057-100MG A1541057-1G A1541057-250MG
76668-324EA 37.14 USD
76668-324 76668-326 76668-328
2-(2-Fluoro-3-isobutoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%
2-(2-Fluoro-3-isobutoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₆H₂₄BFO₃
MW: 294.16 g/mol
CAS Number: 2828440-13-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR