(3,5-Diamino-4-isobutylphenyl)boronic acid 95%

Supplier: Ambeed
Warning

A1540325-100MG A1540325-1G A1540325-250MG
76668-312EA 37.14 USD
76668-312 76668-314 76668-316
(3,5-Diamino-4-isobutylphenyl)boronic acid 95%
(3,5-Diamino-4-isobutylphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₇BN₂O₂
MW: 208.06 g/mol
CAS Number: 2828440-12-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR