2-(4-((3-Chlorobenzyl)oxy)-3,5-dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%
Supplier: Ambeed
|
Warning
|
A1540255-100MG
A1540255-1G
A1540255-250MG
76668-298EA
37.14
USD
76668-298
76668-300
76668-302
2-(4-((3-Chlorobenzyl)oxy)-3,5-dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%
2-(4-((3-Chlorobenzyl)oxy)-3,5-dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Precaución: for Research Use Only
Formula:
C₂₁H₂₆BClO₃ MW: 372.69 g/mol |
CAS Number:
2828440-10-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...