2-(4'-(Isopentyloxy)-[1,1'-biphenyl]-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%

Supplier: Ambeed
Warning

A1539996-100MG A1539996-1G A1539996-250MG
76668-292EA 37.14 USD
76668-292 76668-294 76668-296
2-(4'-(Isopentyloxy)-[1,1'-biphenyl]-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%
2-(4'-(Isopentyloxy)-[1,1'-biphenyl]-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₂₃H₃₁BO₃
MW: 366.30 g/mol
CAS Number: 2828440-09-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR