tert-Butyl (2-((4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)sulfonamido)ethyl)carbamate 95%

Supplier: Ambeed
Warning

A1539976-100MG A1539976-1G A1539976-250MG
76668-286EA 19 USD
76668-286 76668-288 76668-290
tert-Butyl (2-((4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)sulfonamido)ethyl)carbamate 95%
tert-Butyl (2-((4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)sulfonamido)ethyl)carbamate

Precaución: for Research Use Only

Formula: C₁₉H₃₁BN₂O₆S
MDL Number: MFCD28128481
CAS Number: 1014613-14-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR