tert-Butyl ((3-azabicyclo[3.1.0]hexan-6-yl)methyl)(methyl)carbamate 95%

Supplier: Ambeed
Warning

A1539329-1G A1539329-100MG A1539329-250MG
76668-276EA 395 USD
76668-276 77876-229 77876-230
tert-Butyl ((3-azabicyclo[3.1.0]hexan-6-yl)methyl)(methyl)carbamate 95%
tert-Butyl ((3-azabicyclo[3.1.0]hexan-6-yl)methyl)(methyl)carbamate

Precaución: for Research Use Only

Formula: C₁₂H₂₂N₂O₂
MDL Number: MFCD28634726
CAS Number: 1781667-67-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR