4-Bromo-4'-(isopentyloxy)-1,1'-biphenyl 95+%

Supplier: Ambeed
Warning

A1539265-100MG A1539265-1G A1539265-250MG
76668-270EA 36.08 USD
76668-270 76668-272 76668-274
4-Bromo-4'-(isopentyloxy)-1,1'-biphenyl 95+%
4-Bromo-4'-(isopentyloxy)-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₇H₁₉BrO
MW: 319.23 g/mol
CAS Number: 2803477-16-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR