Dimethyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)isophthalate 95+%

Supplier: Ambeed
Warning

A1539259-100MG A1539259-1G A1539259-250MG A1539259-5G
76668-256EA 22 USD
76668-256 76668-258 76668-260 76668-262
Dimethyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)isophthalate 95+%
Dimethyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)isophthalate

Precaución: for Research Use Only

Formula: C₁₆H₂₁BO₆
MDL Number: MFCD23379495
CAS Number: 1186376-95-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR