(4-Fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)(morpholino)methanone 97%
Supplier: Ambeed
|
Warning
|
A1539028-100MG
A1539028-1G
A1539028-250MG
76668-244EA
37.14
USD
76668-244
76668-246
76668-248
(4-Fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)(morpholino)methanone 97%
(4-Fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)(morpholino)methanone
Precaución: for Research Use Only
Formula:
C₁₇H₂₃BFNO₄ MW: 335.17 g/mol |
CAS Number:
2828440-06-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...