2,6-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde 95%

Supplier: Ambeed
Warning

A1539016-100MG A1539016-1G A1539016-250MG
76668-232EA 35.01 USD
76668-232 76668-234 76668-236
2,6-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde 95%
2,6-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₅H₂₁BO₃
MW: 260.13 g/mol
MDL Number: MFCD32902841
CAS Number: 2828440-05-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR