(3-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanol 95+%

Supplier: Ambeed
Warning

Synonyms: 2-Methyl-4-(hydroxymethyl)phenylboronic acid pinacol ester

A1538886-100MG A1538886-1G A1538886-250MG
76668-226EA 37.14 USD
76668-226 76668-228 76668-230
(3-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanol 95+%
(3-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanol

Precaución: for Research Use Only

Formula: C₁₄H₂₁BO₃
MW: 248.12 g/mol
MDL Number: MFCD18732621
CAS Number: 1370732-71-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR