(3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)(thiomorpholino)methanone 97%
Supplier: Ambeed
|
Warning
|
A1538722-100MG
A1538722-1G
A1538722-250MG
76668-214EA
36.08
USD
76668-214
76668-216
76668-218
(3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)(thiomorpholino)methanone 97%
(3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)(thiomorpholino)methanone
Precaución: for Research Use Only
Formula:
C₁₇H₂₄BNO₃S MW: 333.25 g/mol |
MDL Number:
MFCD28167020 CAS Number: 1509931-83-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...