Ethyl 4-ethoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate 95+%

Supplier: Ambeed
Warning

A1538299-100MG A1538299-1G A1538299-250MG
76668-196EA 19 USD
76668-196 76668-198 76668-200
Ethyl 4-ethoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate 95+%
Ethyl 4-ethoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

Precaución: for Research Use Only

Formula: C₁₇H₂₅BO₅
CAS Number: 2828439-80-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR