N-(4,5-Dihydrothiazol-2-yl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide 95%
Supplier: Ambeed
|
Warning
|
A1537616-100MG
A1537616-1G
A1537616-250MG
76668-160EA
19
USD
76668-160
76668-162
76668-164
N-(4,5-Dihydrothiazol-2-yl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide 95%
N-(4,5-Dihydrothiazol-2-yl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
Precaución: for Research Use Only
Formula:
C₁₆H₂₁BN₂O₃S |
CAS Number:
2828439-73-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...