(3,4-Dichloro-5-methoxyphenyl)boronic acid 95%

Supplier: Ambeed
Warning

A1537614-100MG A1537614-1G A1537614-250MG
76668-154EA 37 USD
76668-154 76668-156 76668-158
(3,4-Dichloro-5-methoxyphenyl)boronic acid 95%
(3,4-Dichloro-5-methoxyphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₇BCl₂O₃
MDL Number: MFCD31622685
CAS Number: 2828439-72-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR