(3-Methyl-4-nitrophenyl)boronic acid 95%

Supplier: Ambeed
Warning

A1537283-100MG A1537283-1G A1537283-250MG A1537283-5G
76668-148EA 44 USD
76668-148 76668-150 76668-152 77876-210
(3-Methyl-4-nitrophenyl)boronic acid 95%
(3-Methyl-4-nitrophenyl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₈BNO₄
MDL Number: MFCD28147723
CAS Number: 1436619-60-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR