2-(5-Chloropyridin-2-yl)-6-methyl-1,3,6,2-dioxazaborocane-4,8-dione 95%

Supplier: Ambeed
Warning

A1537265-100MG A1537265-250MG
76668-144EA 36.08 USD
76668-144 76668-146
2-(5-Chloropyridin-2-yl)-6-methyl-1,3,6,2-dioxazaborocane-4,8-dione 95%
2-(5-Chloropyridin-2-yl)-6-methyl-1,3,6,2-dioxazaborocane-4,8-dione

Precaución: for Research Use Only

Formula: C₁₀H₁₀BClN₂O₄
MW: 268.46 g/mol
CAS Number: 2639478-54-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR