(3-Bromo-5-(cyclopropylmethoxy)phenyl)boronic acid 95%

Supplier: Ambeed
Warning

A1537161-100MG A1537161-1G A1537161-250MG
76668-126EA 36.08 USD
76668-126 76668-128 76668-130
(3-Bromo-5-(cyclopropylmethoxy)phenyl)boronic acid 95%
(3-Bromo-5-(cyclopropylmethoxy)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₂BBrO₃
MW: 270.91 g/mol
CAS Number: 2828439-70-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR