(4-(Methoxycarbonyl)-1H-indol-6-yl)boronic acid 95%

Supplier: Ambeed
Warning

A1536411-100MG A1536411-1G A1536411-250MG
76668-118EA 37 USD
76668-118 76668-120 76668-122
(4-(Methoxycarbonyl)-1H-indol-6-yl)boronic acid 95%
(4-(Methoxycarbonyl)-1H-indol-6-yl)boronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀BNO₄
MW: 219.00 g/mol
CAS Number: 2828439-69-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR