(3-Bromo-4,5-dimethylphenyl)boronic acid 95+%

Supplier: Ambeed
Warning

A1536329-100MG A1536329-1G A1536329-250MG
76668-106EA 19 USD
76668-106 76668-108 76668-110
(3-Bromo-4,5-dimethylphenyl)boronic acid 95+%
(3-Bromo-4,5-dimethylphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₁₀BBrO₂
CAS Number: 2828439-67-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR